Learn More
Sino Biological Anti-MKK6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100954T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MKK6 / MEK6 / MAP2K6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2472