Learn More
Sino Biological Anti-MKK7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100955T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Map2k7 / MKK7, and purified by antigen affinity chromatography.
Catalog No. 50-161-2475