Learn More
Sino Biological Anti-p38 beta/MAPK11 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100956T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MAPK11, and purified by antigen affinity chromatography.
Catalog No. 50-161-2481