Learn More
Sino Biological Anti-JNK2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100961T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human JNK2/MAPK9, and purified by antigen affinity chromatography.
Catalog No. 50-161-2493