Learn More
Sino Biological Anti-MEIS2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100968T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MEIS2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2505