Learn More
Sino Biological Recombinant Anti-MSH6 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 100973R035100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human MSH6 / GTBP. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-2517