Learn More
Sino Biological Anti-MUSK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100975T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MUSK, and purified by antigen affinity chromatography.
Catalog No. 50-161-2528