Learn More
Sino Biological Recombinant Human IL-8/CXCL8 Protein (aa 28-99, hFc Tag)
Supplier:  Sino Biological 10098H01H2100
Encompass_Preferred
A DNA sequence encoding the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3983