Learn More
Sino Biological Recombinant Human IL-8/CXCL8 Protein (His Tag)
Supplier:  Sino Biological 10098H08Y100
Encompass_Preferred
A DNA sequence encoding the human CXCL8 (NP_000575.1) (Ser28-Ser99) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3985