Learn More
Sino Biological Anti-PAK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100983T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human PAK1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2531