Learn More
Sino Biological Anti-PARP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100984T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PARP-1 / PARP, and purified by antigen affinity chromatography.
Catalog No. 50-161-2534