Learn More
Sino Biological Recombinant Human S6K1/RPS6KB1 Protein (GST Tag)
Supplier:  Sino Biological 10099H09B50
Encompass_Preferred
A DNA sequence encoding the human RPS6KB1 (P23443-Alpha I) (Met1-Leu525) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-3994