Learn More
Sino Biological Anti-S6K1/RPS6KB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10099T38100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PS6K / RPS6KB1 (rh PS6K / RPS6KB1; Catalog#10099-H09B; P23443; Met1-Leu525). PS6K / RPS6KB1 specific IgG was purified by Human PS6K / RPS6KB1 affinity chromatography.
Catalog No. 50-161-2581