Learn More
Sino Biological Anti-PML Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100997T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PML / RNF71 / TRIM19, and purified by antigen affinity chromatography.
Catalog No. 50-161-2569