Learn More
LAMDA BIOTECH INC 100bp DNA Ladder
Catalog No. 502045810
Encompass_Preferred
100bp DNA Ladder consists of 11 DNA fragments ranging in size from 100-1500 base pairs (bp). 6µl will yield at least 30ng DNA in any single band. The intensity of the 500bp band has been increased to serve as a reference for easy identification. . This ladder can also be used side by side with the 100bp-50 DNA Ladder (Cat. No. M109) for more accurate estimation of the size your DNA fragments.
Catalog No. 50-204-5810 Supplier LAMDA BIOTECH INC Supplier No. M107
100bp DNA Ladder consists of 11 DNA fragments ranging in size from 100-1500 base pairs (bp). 6µl will yield at least 30ng DNA in any single band. The intensity of the 500bp band has been increased to serve as a reference for easy identification. . This ladder can also be used side by side with the 100bp-50 DNA Ladder (Cat. No. M109) for more accurate estimation of the size your DNA fragments.