Learn More
LAMDA BIOTECH INC 100bp Plus DNA Ladder
Catalog No. NC3726409
Encompass_Preferred
A molecular weight marker containing 13 distinct DNA fragments ranging from 100-3000 base pairs (bp), featuring enhanced 500 bp and 1500 bp reference bands for easy orientation.
Catalog No. NC3726409 Supplier LAMDA BIOTECH INC Supplier No. M111
A molecular weight marker containing 13 distinct DNA fragments ranging from 100-3000 base pairs (bp), featuring enhanced 500 bp and 1500 bp reference bands for easy orientation.