Learn More
Taylor Wharton 100CELL SS RACK K&LABS SERIES
Catalog No. NC9102500
Encompass_Preferred
Inventory Rack for storage of 100 Vials - 13 shelves tall for 2 in. (51mm) boxes.
Catalog No. NC9102500 Supplier Taylor Wharton Supplier No. R23K8C35
Inventory Rack for storage of 100 Vials - 13 shelves tall for 2 in. (51mm) boxes.