Learn More
Brookfield Engineer Labs Brookfield Ametek Mineral Standard 100 CPS at 25C 46 CPS at40C S60
Catalog No. NC0780806
Encompass
Silicone viscosity standard fluids are most commonly used to verify calibration of Brookfield lab viscometers/ rheometers and are less temperature sensitive than mineral oils.
Catalog No. NC0780806 Supplier Brookfield Engineer Labs Supplier No. 100 CPS AT 25C
Silicone viscosity standard fluids are most commonly used to verify calibration of Brookfield lab viscometers/ rheometers and are less temperature sensitive than mineral oils.