Learn More
Trajan Scientific 100F-CTC-GT-5/0.47C
SDP
Catalog No. 502530327
Encompass_Preferred
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 100 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Length Needle: 50 mm, Needle Gauge: 26 G, Needle Point Style: Cone, Description: Gas tight, Diameter Outer Needle: 0.63 mm
Catalog No. 50-253-0327 Supplier Trajan Scientific Supplier No. 005700
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 100 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Length Needle: 50 mm, Needle Gauge: 26 G, Needle Point Style: Cone, Description: Gas tight, Diameter Outer Needle: 0.63 mm