Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 100g, 1-Boc-3,6-Dihydro-2H-pyridine-4-boronic acid pinacol ester, 98%, [286961-14-6], P1164 309.209 C16H28BNO4

Catalog No. 5038858
Encompass_Preferred

Compound: 1-Boc-3,6-Dihydro-2H-pyridine-4-boronic acid pinacol ester

CAS#: 286961-14-6

MDL#: MFCD03840345

Background: A Boc-protected tetrahydropyridine boronic ester widely used in Suzuki-Miyaura cross-coupling reactions in organic and pharmaceutical synthesis. It is employed in the preparation of enzymatic inhibitors and receptor ligands in drug discovery research.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-388-58 Supplier Frontier Specialty Chemicals Supplier No. P1164100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Boc-3,6-Dihydro-2H-pyridine-4-boronic acid pinacol ester

CAS#: 286961-14-6

MDL#: MFCD03840345

Background: A Boc-protected tetrahydropyridine boronic ester widely used in Suzuki-Miyaura cross-coupling reactions in organic and pharmaceutical synthesis. It is employed in the preparation of enzymatic inhibitors and receptor ligands in drug discovery research.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.