Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 100g, 4-(Methoxycarbonyl)phenylboronic acid, 97%, [99768-12-4], M3563 179.97 C8H9BO4

Catalog No. 5038489
Encompass_Preferred

Compound: 4-(Methoxycarbonyl)phenylboronic acid

CAS#: 99768-12-4

MDL#: not listed

Background: A methyl ester-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and materials science research. The ester group provides a handle for later hydrolysis or amidation.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-384-89 Supplier Frontier Specialty Chemicals Supplier No. M3563100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(Methoxycarbonyl)phenylboronic acid

CAS#: 99768-12-4

MDL#: not listed

Background: A methyl ester-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical and materials science research. The ester group provides a handle for later hydrolysis or amidation.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.