Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 100g, 5-Bromoindole, 99%, [10075-50-0], B1579 196.05 C8H6BrN

Catalog No. 50981282
Encompass_Preferred

Compound: 5-Bromoindole

CAS#: 10075-50-0

MDL#: MFCD00005670

Background: 5-Bromoindole is a crucial building block in the synthesis of pharmaceuticals targeting neurological disorders, cardiovascular conditions, tumors, and immune function. It is a potential inhibitor of GSK-3 and serves as a probe in biochemical studies of protein-ligand interactions, as well as a building block for fluorescent imaging probes and organic electronic materials.

Catalog No. 50-981-282 Supplier Frontier Specialty Chemicals Supplier No. B1579100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromoindole

CAS#: 10075-50-0

MDL#: MFCD00005670

Background: 5-Bromoindole is a crucial building block in the synthesis of pharmaceuticals targeting neurological disorders, cardiovascular conditions, tumors, and immune function. It is a potential inhibitor of GSK-3 and serves as a probe in biochemical studies of protein-ligand interactions, as well as a building block for fluorescent imaging probes and organic electronic materials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.