Learn More
The Fresh Test 100g Glucose Bottle 10/pack, Non-GMO Dextrose
Catalog No. NC3627364 Shop All The Fresh Test Diagnostic Glucose Test Products
Encompass
The Fresh Test 100g Glucose Bottle is a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 100 grams of glucose in liquid form. For an optimal experience, the beverage should be consumed cold. When used for a 100-gram glucose load test, the product should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 10 units per box.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
Catalog No. NC3627364 Supplier The Fresh Test Supplier No. 2710205910BOX
The Fresh Test 100g Glucose Bottle is a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 100 grams of glucose in liquid form. For an optimal experience, the beverage should be consumed cold. When used for a 100-gram glucose load test, the product should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 10 units per box.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.