Learn More
The Fresh Test 100g Glucose Powder 10 packs, Non-GMO Dextrose
Catalog No. NC3627358 Shop All The Fresh Test Diagnostic Glucose Test Products
Encompass
The Fresh Test 100g Glucose Powder is a diagnostic glucose beverage intended for use in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each pouch contains 100 grams of glucose to be reconstituted with 10 oz. of water prior to administration. For an optimal experience, the beverage should be consumed cold. When used for a 100-gram glucose load test, the beverage should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free pouch
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 10 units per box.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
Catalog No. NC3627358 Supplier The Fresh Test Supplier No. 2710205710BOX
The Fresh Test 100g Glucose Powder is a diagnostic glucose beverage intended for use in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each pouch contains 100 grams of glucose to be reconstituted with 10 oz. of water prior to administration. For an optimal experience, the beverage should be consumed cold. When used for a 100-gram glucose load test, the beverage should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free pouch
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Ordering Information
This catalog number represents 10 units per box.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.