Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptomycin sulfate salt, 3810-74-0, MFCD00037023, 100 g
Catalog No. NC0809046
Encompass
Linear Formula: C21H39N7O12 · 1.5 H2SO4, Molecular Weight: 728.69, powder, color: white to off-white.
Catalog No. NC0809046 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S6501100G
Linear Formula: C21H39N7O12 · 1.5 H2SO4, Molecular Weight: 728.69, powder, color: white to off-white.