Learn More
Chemglass Life Sciences 100L Jacketed Reaction Vessel, Large Scale, 3" Beaded Pipe Drain Valve, 400mm Flange, Squatty
SDP
Catalog No. 501531335
Encompass_Preferred
100L Jacketed Reaction Vessel, Large Scale, 3" Beaded Pipe Drain Valve, 400mm Flange, Squatty. Jacketed large scale process reaction vessels. The 100L vessels have a 400mm ID flat ground flange. The inlet/outlet of the vessels are beaded pipe fittings for use with the CG-1964 and CG-1969-A beaded pipe adapters. Reactors have a 3 beaded pipe valve that is detachable. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. The 100L vessels equipped with the detachable zero dead space valve have a lower silicone seal. Vessels are serial numbered and have a graduation scale on both sides showing approximate volume. PLEASE NOTE: BOTTOM DRAIN VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.
Catalog No. 50-153-1335 Supplier Chemglass Life Sciences Supplier No. CG1968W101
Add to Cart
Add to Cart
Request a Quote
100L Jacketed Reaction Vessel, Large Scale, 3" Beaded Pipe Drain Valve, 400mm Flange, Squatty. Jacketed large scale process reaction vessels. The 100L vessels have a 400mm ID flat ground flange. The inlet/outlet of the vessels are beaded pipe fittings for use with the CG-1964 and CG-1969-A beaded pipe adapters. Reactors have a 3 beaded pipe valve that is detachable. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. The 100L vessels equipped with the detachable zero dead space valve have a lower silicone seal. Vessels are serial numbered and have a graduation scale on both sides showing approximate volume. PLEASE NOTE: BOTTOM DRAIN VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.