Learn More
Chemglass Life Sciences Reactor System, 100L Tall, Complete, Jacketed, 400mm Flange, Electric Motor, with Manifold System
SDP
Catalog No. 501531336
Encompass_Preferred
Reactor System, 100L Tall, Complete, Jacketed, 400mm Flange, Electric Motor, with Manifold System. 100L Tall glass reactor system with 400mm flange on Chemglass's innovative Kwik Mount support frame. Lexan safety shields surround the reactor with the front panel hinged for easy access. 400mm ID Flange Provides Wider Access For Insertion or Probes, Baffles and Other Accessories without Interfering with Agitators. Heavy Duty 2" Diameter Solid Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Electric and XP Motor has Separate Controller with Digital Display. Reactor Jacket Graduation Scale on Both Sides.
Catalog No. 50-153-1336 Supplier Chemglass Life Sciences Supplier No. CG1968W6100
Add to Cart
Add to Cart
Request a Quote
Reactor System, 100L Tall, Complete, Jacketed, 400mm Flange, Electric Motor, with Manifold System. 100L Tall glass reactor system with 400mm flange on Chemglass's innovative Kwik Mount support frame. Lexan safety shields surround the reactor with the front panel hinged for easy access. 400mm ID Flange Provides Wider Access For Insertion or Probes, Baffles and Other Accessories without Interfering with Agitators. Heavy Duty 2" Diameter Solid Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Electric and XP Motor has Separate Controller with Digital Display. Reactor Jacket Graduation Scale on Both Sides.