Learn More
Chemglass Life Sciences Shielded Reactor Support Frame, Mobile, Jacketed, 100L, 400mm Flange, 29.75" W x 25.0" D x 83.5" H
SDP
Catalog No. 501531344
Encompass_Preferred
Shielded Reactor Support Frame, Mobile, Jacketed, 100L, 400mm Flange. These innovative Kwik Mount reactor support frame is designed to be user friendly. Motor mount arm can be used with either electric, air, or XP motor. Reactors and glassware can be quickly installed and removed saving time and frustration normally associated with out-of-date designs. Support frame is to be used for 100L 400mm flange Jacketed reactor systems and is supplied with Lexan safety shields.
Catalog No. 50-153-1344 Supplier Chemglass Life Sciences Supplier No. CG1968Y24M
Add to Cart
Add to Cart
Request a Quote
Shielded Reactor Support Frame, Mobile, Jacketed, 100L, 400mm Flange. These innovative Kwik Mount reactor support frame is designed to be user friendly. Motor mount arm can be used with either electric, air, or XP motor. Reactors and glassware can be quickly installed and removed saving time and frustration normally associated with out-of-date designs. Support frame is to be used for 100L 400mm flange Jacketed reactor systems and is supplied with Lexan safety shields.