Learn More
Bio X Cell Antibody, Anti-Mouse IL-12 p40, 100 MG
Supplier: Bio X Cell BP0051R100MG
The C17.8 antibody reacts with mouse p40, also known as IL-12β. This 40 kDa subunit is part of IL-12 and IL-23. The antibody neutralizes both IL-12 and IL-23 bioactivity.
- Validated for in vivo IL-12p40 neutralization
- Suitable for p40 affinity chromatography
- Applications include Immunoprecipitation, ELISA, Flow cytometry, and Western blot
- Formulated in PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin level ≤0.5EU/mg
- Aggregation less than 5%
- Purity ≥95%
- Sterility achieved by 0.2 µm filtration
- Produced in an animal-free facility and purified by Protein G
- Molecular weight is 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.