Learn More
SPECTRACER CPACHEM LLC 100MG FATTYACID MIX ACC TO PH
Supplier:  SPECTRACER CPACHEM LLC CM50415
Encompass_Preferred
100mg Fatty Acid Mix acc to Ph Eur 2.4.22-3 CRM, ISO 17034 and ISO 17025-Shelf Life 24mos, Store in a freezer at -18degrees Celsius or below.
Catalog No. NC1912353