Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Denatonium Benzoate, Anhydrous, Granular, NF, 99.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18602043 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-043 5 kg Amber Glass Bottle
18-602-040 5 g Amber Glass Bottle
18-602-039 1 g Amber Glass Bottle
18-602-041 25 g Amber Glass Bottle
18-602-042 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-043 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE1015KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

DE101, 3734-33-6, C28H34N2O3

Spectrum™ Chemical Denatonium Benzoate, Anhydrous, Granular, NF is used as a denaturant and masking agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3734-33-6
Molecular Formula C28H34N2O3
Molecular Weight (g/mol) 446.59
MDL Number MFCD00031578
InChI Key VWTINHYPRWEBQY-UHFFFAOYSA-N
IUPAC Name benzyl({[(2,6-dimethylphenyl)carbamoyl]methyl})diethylazanium benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CC[N+](CC)(CC(=O)NC1=C(C)C=CC=C1C)CC1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Melting Range 163°C to 170°C
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 99.5 to 101%
Grade NF
Physical Form Granules
Chemical Name or Material Denatonium Benzoate, Anhydrous, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.