Learn More
GLPBIO TECHNOLOGY INC 100mg H-Gln-AMCFluorogenic substrate used in a continuous spectrometric assay for glutaminyl cyclase activity
Catalog No. NC3368117
Encompass_Preferred
100mg H-Gln-AMCFluorogenic substrate used in a continuous spectrometric assay for glutaminyl cyclase activity
Catalog No. NC3368117 Supplier GLPBIO TECHNOLOGY INC Supplier No. GA22334100MG
100mg H-Gln-AMCFluorogenic substrate used in a continuous spectrometric assay for glutaminyl cyclase activity