Learn More
Quality Biological Inc Ampicillin Ready-Made Solution (1mL), Pack of 10 X 1 mL
SDP
Catalog No. 502209781
Encompass_Preferred
Ampicillin in a convenient ready-to-use solution useful in selecting for ampicillin resistant bacteria. Ampicillin is a beta-lactam antibiotic with an amino group side chain attached to the penicillin structure.
Convenient 1000X concentrated stock
Small 1 ml size to prevent freeze-thaw cycle
Affordable price
Manufactured in an ISO 9001:2015 certified facility.
Catalog No. 50-220-9781 Supplier Quality Biological Inc Supplier No. 351344731
Ampicillin in a convenient ready-to-use solution useful in selecting for ampicillin resistant bacteria. Ampicillin is a beta-lactam antibiotic with an amino group side chain attached to the penicillin structure.
Convenient 1000X concentrated stock
Small 1 ml size to prevent freeze-thaw cycle
Affordable price
Manufactured in an ISO 9001:2015 certified facility.