Learn More
Teknova 100mg/mL Carbenicillin.10mL Tubes, 25-Pack, Sterile. 4 pack.
SDP
Catalog No. 502387279
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-7279 Supplier Teknova Supplier No. C21314PK
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.