Learn More
Teknova 100mg/mL Carbenicillin. 25mL, Sterile
SDP
Catalog No. NC2398932
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. NC2398932 Supplier Teknova Supplier No. C2130
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.