Learn More
Teknova 100mg/mL Carbenicillin Solution. 1mL Tubes, Sterile, 50 Tubes per Rack.
SDP
Catalog No. 502388175
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8175 Supplier Teknova Supplier No. C2136
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.