Learn More
Teknova 100mg/mL Kanamycin Solution. 1mL, 100-Pack, Sterile. 2 pack.
SDP
Catalog No. 502387409
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-7409 Supplier Teknova Supplier No. K21352PK
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.