Learn More
Teknova 100mg/mL Spectinomycin Solution. 1mL Tube, 50-Pack, Sterile.
SDP
Catalog No. 502388166
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8166 Supplier Teknova Supplier No. S8001
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.