Learn More
Ops Diagnostics 100 micron Zirconium Beads, 1200 mg Pre-Filled Tubes (100 count)
Supplier:  Ops Diagnostics PFAW 10010026
Encompass
Tougher than silica, less expensive than stainless steel, and inert in most solutions, 100 µm Acid Washed Zirconium Beads are suitable for homogenizing bacteria and small tissues. May also be used as a sonication additive.Beads are available in Acid Washed, Low Binding, and and Molecular Biology Grade format. Since untreated beads are notorious for containing large amounts of debris and dust, all beads are acid washed, at a minimum, prior to packaging. Low Binding Beads are chemically altered using a proprietary process to bind less biomolecules liberated from homogenized samples. Pre-Filled Tubes are a convenient solution for labs seeking time savings by no longer requiring researchers to weigh, fill and QC individual tubes. They are available in 2 mL skirted (i.e., self-standing) or unskirted polypropylene tubes to ensure compatibility with most tube homogenizers. Clear screw caps with O-rings included.
Catalog No. NC1419519