Learn More
METTLER TOLEDO Mettler Toledo 100 mL Glass Reactor Set, One-Piece Design, 5 Ports, Magnetic & Overhead Stirring
Catalog No. NC1434254 Shop All METTLER TOLEDO Products
Encompass
The METTLER TOLEDO One-Piece Reactor Set is engineered for efficient chemical synthesis, offering enhanced temperature control and compatibility with standard lab glassware. Its versatile design supports both magnetic and overhead stirring, making it ideal for various reaction types. With a maximum working volume of 100 mL, this reactor improves efficiency in research projects while meeting traditional flask requirements, differentiating itself with its quick setup and ease of handling.
Catalog No. NC1434254 Supplier METTLER TOLEDO Supplier No. 51161805
The METTLER TOLEDO One-Piece Reactor Set is engineered for efficient chemical synthesis, offering enhanced temperature control and compatibility with standard lab glassware. Its versatile design supports both magnetic and overhead stirring, making it ideal for various reaction types. With a maximum working volume of 100 mL, this reactor improves efficiency in research projects while meeting traditional flask requirements, differentiating itself with its quick setup and ease of handling.