Learn More
SPEX CERTIPREP LLC 3P 100ML 2% HNO3 + TR HF ICP-MS
Supplier: SPEX CERTIPREP LLC 3P ICPMSCSM100
(HPS by Zeptometrix) 32 element calibration standard in 2% HNO3 + Tr HF, Al, Sb, As, Ba, Be, Bi, B, Cd, Ca, Cr, Cu, Co, Eu, Ho, La, Pb, Li, MG, Mn, Mo, Ni, FE, Se, Ag, Na, Sr, Tl, Th, V, U, Yb, Zn at 10 ppm 1 year expiry date
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.