Learn More
Chemglass Life Sciences Chemglass 100 mL 2-Neck Pear-Shaped Heavy-Wall Flask, 14/20 Joints (CG-1558-18)
SDP
Catalog No. 502733581
Encompass_Preferred
100 mL heavy-wall, pear-shaped two-neck flask with a center neck and an angled side neck (20°). Both necks use standard 14/20 outer joints. The flask is intended for small-scale reaction, distillation, and reflux applications where an angled side neck improves access for condensers, thermometers, or addition funnels.
  • Heavy-wall pear-shaped body for improved mechanical strength and even heating.
  • Two-neck configuration with one center neck and one side neck angled 20° for auxiliary attachments.
  • Standard 14/20 outer joints for compatibility with common ground-glass fittings.
  • 100 mL capacity suitable for small-scale synthesis and analytical procedures.
  • Designed for applications such as heating under reflux, distillation, and general reaction work.
Catalog No. 50-273-3581 Supplier Chemglass Life Sciences Supplier No. CG155818
100 mL heavy-wall, pear-shaped two-neck flask with a center neck and an angled side neck (20°). Both necks use standard 14/20 outer joints. The flask is intended for small-scale reaction, distillation, and reflux applications where an angled side neck improves access for condensers, thermometers, or addition funnels.
  • Heavy-wall pear-shaped body for improved mechanical strength and even heating.
  • Two-neck configuration with one center neck and one side neck angled 20° for auxiliary attachments.
  • Standard 14/20 outer joints for compatibility with common ground-glass fittings.
  • 100 mL capacity suitable for small-scale synthesis and analytical procedures.
  • Designed for applications such as heating under reflux, distillation, and general reaction work.