Learn More
Ace Glass, Inc. Trap, rotary evaporator, self-washing, 100mL, 24/40 top, 14/20bottom
SDP
Catalog No. 50973364
Encompass_Preferred
100mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 top joint and 14/20 bottom joint.
Catalog No. 50-973-364 Supplier Ace Glass, Inc. Supplier No. 670305
100mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 top joint and 14/20 bottom joint.