Learn More
Ace Glass, Inc. 100mL 24/40-14/20 trap anti-climb
SDP
Supplier:  Ace Glass, Inc. 670605
Encompass_Preferred
100mL anti-climb rotary evaporator trap has upper tabulation to prevent migration of film and foam directly into the condenser. Trap has 24/40 and 14/20 standard taper joints.
Catalog No. 50-978-863