Learn More
Ace Glass, Inc. Beaker, 100mL, jacketed, Ace-Safe hose connections for 1/4intubing, 61mm inside height, 48mm inside diameter
SDP
Catalog No. 50971847
Encompass_Preferred
100mL jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 1/4 in. tubing. Dimensions: 61mm I.H. x 48mm I.D. Does not have a pouring spout.
Catalog No. 50-971-847 Supplier Ace Glass, Inc. Supplier No. 5340103
Add to Cart
Add to Cart
100mL jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 1/4 in. tubing. Dimensions: 61mm I.H. x 48mm I.D. Does not have a pouring spout.