Learn More
Chemglass Life Sciences 100mL Funnel, Addition, 14/20 Joint Size, 2mm Glass Stpk On Side Arm, Airfree, Schlenk
SDP
Catalog No. AF054801
Encompass_Preferred
100mL Funnel, Addition, 14/20 Joint Size, 2mm Glass Stpk On Side Arm, Airfree, Schlenk. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with the stopcock.
Catalog No. AF054801 Supplier Chemglass Life Sciences Supplier No. AF054801
100mL Funnel, Addition, 14/20 Joint Size, 2mm Glass Stpk On Side Arm, Airfree, Schlenk. Lower outlet is a standard taper inner joint and has a CG-961-01 0-4mm Chem-Vac Chem-Cap valve. Top outer joint is the same size as the lower inner joint. Available with a choice of a 2mm high vacuum glass stopcock having a Stainless Steel constant pressure retaining device or a 0-4mm Chem-Cap Airfree inlet valve. This item comes with the stopcock.