Learn More
Ace Glass, Inc. Mantle, 100mL, for 60mm diameter spherical or cylindrical flask, 60mm ID x 30.2mm height, 80w-115v, CSA rated
SDP
Catalog No. 50977322
Encompass
100mL aluminum housed, table-model mantle for 60mm diameter spherical or cylindrical flask. 60mm I.D. x 30.2mm height. Maximum internal operating temperature is 450 Deg. centigrade. 80w-115v, cULus rated. Power cord included.
Catalog No. 50-977-322 Supplier Ace Glass, Inc. Supplier No. 1205307
100mL aluminum housed, table-model mantle for 60mm diameter spherical or cylindrical flask. 60mm I.D. x 30.2mm height. Maximum internal operating temperature is 450 Deg. centigrade. 80w-115v, cULus rated. Power cord included.