Learn More
Chemglass Life Sciences 100mL Beaker, Low Form, Griffin, Pyrex
SDP
Catalog No. 501214795
Encompass_Preferred
100mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.
Catalog No. 50-121-4795 Supplier Chemglass Life Sciences Supplier No. CG8048P100
100mL Beaker, Low Form, Griffin, Pyrex. Manufactured with uniform wall thickness these beakers offer the optimum balance between thermal shock resistance and mechanical strength. All sizes have an extra large marking spot. Designed from ASTM Specification E960, Type I requirements.