Learn More
US Plastic Corporation 100ML Graduated Beakers, Polypropylene, Disposable
SDP
Catalog No. NC0968026
Encompass
Disposable Polypropylene Beakers, 100ml Graduated Beakers These beakers are low cost, unbreakable and exhibit high corrosion resistance. *Polypropylene *Dripless lip & flat base *Molded-in graduations in ml & oz. *Translucent *Autoclavable *Maximum temperature: 250 Deg.F *Sold in package lots only
Catalog No. NC0968026 Supplier US Plastic Corporation Supplier No. 76054
Disposable Polypropylene Beakers, 100ml Graduated Beakers These beakers are low cost, unbreakable and exhibit high corrosion resistance. *Polypropylene *Dripless lip & flat base *Molded-in graduations in ml & oz. *Translucent *Autoclavable *Maximum temperature: 250 Deg.F *Sold in package lots only