Learn More
Ace Glass, Inc. 100mL graduated buret open at top and bottom
SDP
Catalog No. 50978419
Encompass_Preferred
100mL graduated buret open at top and bottom. Top is beaded, and bottom is fire polished. Used in fabrication of apparatus requiring graduation.
Catalog No. 50-978-419 Supplier Ace Glass, Inc. Supplier No. 575808
100mL graduated buret open at top and bottom. Top is beaded, and bottom is fire polished. Used in fabrication of apparatus requiring graduation.